accessibility menu, dialog, popup

UserName

CAS RN 131790-57-3

CH3OOCH3

CAS RN 131790-57-3

IUPAC Name: 4-(4-pentylcyclohexyl)phenyl 4'-propyl-[1,1'-bi(cyclohexane)]-4-carboxylate
Synonyms: 4-(4-pentylcyclohexyl)phenyl 4'-propyl-[1,1'-bi(cyclohexane)]-4-carboxylate
Molecular Weight (g/mol): 480.78
Molecular Formula: C33H52O2
InChi Key: HNSOCAVSNSWNJJ-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(CC1)C1=CC=C(OC(=O)C2CCC(CC2)C2CCC(CCC)CC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results