CAS RN 13170-43-9
CAS RN 13170-43-9
IUPAC Name:
magnesium(2+) (trimethylsilyl)methanide chloride
Synonyms:
magnesium(2+) (trimethylsilyl)methanide chloride
Molecular Weight (g/mol):
146.97
Molecular Formula:
C4H11ClMgSi
InChi Key:
BXBLTKZWYAHPKM-UHFFFAOYSA-M
SMILES:
[Mg++].[Cl-].C[Si](C)(C)[CH2-]
1
–
2
of
2
results
1
(Trimethylsilyl)methylmagnesium chloride, 1.3M solution in THF, AcroSeal™
CAS: 13170-43-9 Molecular Formula: C4H11ClMgSi Molecular Weight (g/mol): 146.97 MDL Number: MFCD00009916 InChI Key: NAQATMJWCJCHOZ-UHFFFAOYSA-M Synonym: trimethylsilylmethylmagnesium chloride,tmsch2mgcl,trimethylsilyl methylmagnesium chloride,trimethylsilyl methyl magnesium chloride,trimethylsilyl methylmagnesium chloride solution,me3sich2mgcl,naqatmjwcjchoz-uhfffaoysa-m,chlorotrimethylsilylmethyl magnesium,trimethylsiylmethylmagnesium chloride PubChem CID: 2734900 SMILES: C[Si](C)(C)C[Mg]Cl