CAS RN 1313730-08-3
CAS RN 1313730-08-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bisphenol A-13C12 Beta-D-Glucuronide, TRC
CAS: 1313730-08-3 Molecular Formula: 13C12 C9 H24 O8 Molecular Weight (g/mol): 416.32 Synonym: 4-[1-(4-Hydroxyphenyl-13C6)-1-methylethyl]phenyl-13C6 beta-D-glucopyranosiduronic acid,Bisphenol A beta-D-Glucuronide (diphenylen-13C12),Bisphenol A beta-D-Monoglucuronide (diphenylen-13C12) IUPAC Name: (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[4-[2-(4-hydroxy(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl)propan-2-yl](1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl]oxyoxane-2-carboxylic acid SMILES: CC(C)([13c]1[13cH][13cH][13c](O)[13cH][13cH]1)[13c]2[13cH][13cH][13c](O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)[13cH][13cH]2