CAS RN 1310-03-8
IUPAC Name:
nonakis(λ²-lead(2+)) hexakis(fluorosilanetris(olate)) dihydrate
Synonyms:
nonakis(λ²-lead(2+)) hexakis(fluorosilanetris(olate)) dihydrate
Molecular Weight (g/mol):
2471.3
Molecular Formula:
F6H4O20Pb9Si6
InChi Key:
UCXBPZOKVHNQLY-UHFFFAOYSA-N
SMILES:
O.O.[Pb++].[Pb++].[Pb++].[Pb++].[Pb++].[Pb++].[Pb++].[Pb++].[Pb++].[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F
1
–
1
of
1
results
1
Lead(II) hexafluorosilicate dihydrate, Thermo Scientific™
CAS: 1310-03-8 Molecular Formula: F6H4O20Pb9Si6 Molecular Weight (g/mol): 2471.312 MDL Number: MFCD00049509 InChI Key: UCXBPZOKVHNQLY-UHFFFAOYSA-N Synonym: acmc-20aknf,fluoro trioxido silane;lead 2+ ;dihydrate,silicate 2-,hexafluoro-, lead 2+ 1:1 , dihydrate 9ci PubChem CID: 57347846 IUPAC Name: fluoro(trioxido)silane;lead(2+);dihydrate SMILES: O.O.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[O-][Si]([O-])([O-])F.[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2].[Pb+2]