CAS RN 130357-25-4
CAS RN 130357-25-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MedChemExpress Exendin-3
MedChemExpress Exendin-3 is a biologically active peptides isolated from venoms of the Gila monster lizards, Heloderma horridurn.
2
MedChemExpress Exendin-3
MedChemExpress Exendin-3 is a biologically active peptides isolated from venoms of the Gila monster lizards, Heloderma horridurn.