CAS RN 13002-81-8
CAS RN 13002-81-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(Methylsulfanyl)-10-[2-[(2RS)-piperidin-2-yl]ethyl]-10H-phenothiazine Hydrochloride (N-Desmethylthioridazine Hydrochloride; Northioridazine Hydrochloride), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.