CAS RN 129752-86-9
CAS RN 129752-86-9
IUPAC Name:
magnesium(2+) (2,4-dichlorophenyl)methanide chloride
Synonyms:
magnesium(2+) (2,4-dichlorophenyl)methanide chloride
Molecular Weight (g/mol):
219.77
Molecular Formula:
C7H5Cl3Mg
InChi Key:
GVZMYROGYRWDEM-UHFFFAOYSA-M
SMILES:
[Mg++].[Cl-].[CH2-]C1=C(Cl)C=C(Cl)C=C1