accessibility menu, dialog, popup

UserName

CAS RN 1293284-58-8

BOOH3CCH3H3CH3CFOOCH3

CAS RN 1293284-58-8

IUPAC Name: methyl 5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Synonyms: methyl 5-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Molecular Weight (g/mol): 280.1
Molecular Formula: C14H18BFO4
InChi Key: DCNAAOVAUVQRQD-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(C=CC(F)=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results