CAS RN 1286458-00-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fexofenadine-d6 Methyl Ester, TRC
CAS: 1286458-00-1 Molecular Formula: C33H35D6NO4 Molecular Weight (g/mol): 521.72 Synonym: 4-[1-Hydroxy-4-[4-(hydroxydiphenylmethyl)-1-piperidinyl]butyl]-α,α-(dimethyl-d6)benzeneacetic Acid Methyl Ester,Methyl 4-[4-[4-(Hydroxydiphenylmethyl)-1-piperidinyl]-1- hydroxybutyl]-α,α-(dimethyl-d6)phenylacetate SMILES: OC(CCCN1CCC(C(O)(C2=CC=CC=C2)C3=CC=CC=C3)CC1)C4=CC=C(C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C(OC)=O)C=C4