CAS RN 1286165-14-7
CAS RN 1286165-14-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Methylethyl 3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylate (3-Methylflavone-8-carboxylic Acid Isopropyl Ester), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.