accessibility menu, dialog, popup

UserName

CAS RN 128345-57-3

HNNH2

CAS RN 128345-57-3

IUPAC Name: pyrrolidin-3-amine
Synonyms: pyrrolidin-3-amine
Molecular Weight (g/mol): 86.14
Molecular Formula: C4H10N2
InChi Key: NGXSWUFDCSEIOO-UHFFFAOYNA-N
SMILES: NC1CCNC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results