CAS RN 1282514-88-8
CAS RN 1282514-88-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
GDC-0326, MedChemExpress
MedChemExpress GDC-0326 is a potent and selective PI3Kα inhibitor with a Ki of 0.2 nM.