accessibility menu, dialog, popup

UserName

CAS RN 128143-89-5

ClNNN

CAS RN 128143-89-5

IUPAC Name: 4-chloro-6-(pyridin-2-yl)-2,2'-bipyridine
Synonyms: 4-chloro-6-(pyridin-2-yl)-2,2'-bipyridine
Molecular Weight (g/mol): 267.72
Molecular Formula: C15H10ClN3
InChi Key: AHEMFMCEBIJRMU-UHFFFAOYSA-N
SMILES: ClC1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)