CAS RN 1279713-77-7
CAS RN 1279713-77-7
IUPAC Name:
2-[(3-chloro-6-fluoro-1-benzothiophen-2-yl)formamido]-3-phenylpropanoic acid
Synonyms:
2-[(3-chloro-6-fluoro-1-benzothiophen-2-yl)formamido]-3-phenylpropanoic acid
Molecular Weight (g/mol):
377.81
Molecular Formula:
C18H13ClFNO3S
InChi Key:
IAASQMCXDRISAV-UHFFFAOYNA-N
SMILES:
OC(=O)C(CC1=CC=CC=C1)NC(=O)C1=C(Cl)C2=CC=C(F)C=C2S1
1
–
1
of
1
results
1
Tocris Bioscience™ CU CPT 4a
CAS: 1279713-77-7 Molecular Formula: C18H13ClFNO3S