CAS RN 1264754-13-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
PROTAC ERRα ligand 1, MedChemExpress
MedChemExpress PROTAC ERRα ligand 1 is an estrogen-related receptor α (ERRα) antagonist with IC50s of 0.04 and 2.8 μM for ERRα and ERRγ, respectively.