accessibility menu, dialog, popup

UserName

CH3OFFFFNH2

CAS RN 1262198-05-9

IUPAC Name: 2-fluoro-5-methoxy-3-(trifluoromethyl)aniline
Synonyms: 2-fluoro-5-methoxy-3-(trifluoromethyl)aniline
Molecular Weight (g/mol): 209.14
Molecular Formula: C8H7F4NO
InChi Key: HFYMUTVWAXCASY-UHFFFAOYSA-N
SMILES: COC1=CC(N)=C(F)C(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results