accessibility menu, dialog, popup

UserName

ClOFFFOCl

CAS RN 1261779-42-3

IUPAC Name: 4-chloro-2-(trifluoromethoxy)benzoyl chloride
Synonyms: 4-chloro-2-(trifluoromethoxy)benzoyl chloride
Molecular Weight (g/mol): 259.01
Molecular Formula: C8H3Cl2F3O2
InChi Key: ZYZNCCXCNJYIHJ-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=C(C=CC(Cl)=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results