accessibility menu, dialog, popup

UserName

CAS RN 1261573-60-7

CH3OFFFBr

CAS RN 1261573-60-7

IUPAC Name: 2-(bromomethyl)-1-methyl-4-(trifluoromethoxy)benzene
Synonyms: 2-(bromomethyl)-1-methyl-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 269.06
Molecular Formula: C9H8BrF3O
InChi Key: BSYHAIGVGUZQGK-UHFFFAOYSA-N
SMILES: CC1=C(CBr)C=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results