accessibility menu, dialog, popup

UserName

H3COOHOOFFF

CAS RN 1261557-46-3

IUPAC Name: 2-[4-methoxy-3-(trifluoromethoxy)phenyl]acetic acid
Synonyms: [4-methoxy-3-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol): 250.17
Molecular Formula: C10H9F3O4
InChi Key: MMYRFHYSHNVBCU-UHFFFAOYSA-N
SMILES: COC1=C(OC(F)(F)F)C=C(CC(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results