CAS RN 1254472-97-3
CAS RN 1254472-97-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fezagepras sodium, MedChemExpress
MedChemExpress Fezagepras (Setogepram) sodium acts as an orally active agonist for GPR40 and as an antagonist or inverse agonist for GPR84. Fezagepras sodium decreases renal, liver and pancreatic fibrosis. Fezagepras sodium exerts anti-fibrotic, anti-inflammatory and anti-proliferative actions.