CAS RN 125079-83-6
CAS RN 125079-83-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CU-CPT-8m, MedChemExpress
MedChemExpress CU-CPT-8m is a specific TLR8 antagonist, with an IC50 of 67 nM.