CAS RN 124935-88-2
CAS RN 124935-88-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(3RS)-3-(2-Methoxy-5-methylphenyl)-N,N-bis(1-methylethyl)-3-phenylpropan-1-amine ((RS)-Tolterodine Methyl Ether), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.