CAS RN 1247119-36-3
CAS RN 1247119-36-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl (2S)-2-[[[2-(1-Methylethyl)thiazol-4-yl]methyl(methyl)carbamoyl]amino]-4-morpholinobutanoate Oxalate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.