CAS RN 1247119-33-0
CAS RN 1247119-33-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thiazol-5-ylmethyl N-((2R,5R)-5-Amino-1,6-diphenylhexan-2-yl)carbamate Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.