CAS RN 1246833-15-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
10-Hydroxy Amitriptyline Oxalate, TRC
CAS: 1246833-15-7 Molecular Formula: C22H25NO Molecular Weight (g/mol): 383.44 Synonym: 5-[3-(Dimethylamino)propylidene]-10,11-dihydro-5H-dibenzo[a,d]cyclohepten-10-ol Oxalate,Amitriptyline EP Impurity F SMILES: CN(C)CCC=C1C2=CC=CC=C2CC(C3=CC=CC=C31)O.C(=O)(C(=O)O)O