accessibility menu, dialog, popup

UserName

FFFFH2N

CAS RN 123973-22-8

IUPAC Name: 3-fluoro-2-(trifluoromethyl)aniline
Synonyms: 3-fluoro-2-(trifluoromethyl)aniline
Molecular Weight (g/mol): 179.12
Molecular Formula: C7H5F4N
InChi Key: SJMDWKFHBPDEQT-UHFFFAOYSA-N
SMILES: NC1=C(C(F)=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results