accessibility menu, dialog, popup

UserName

NHNFFFFFFI

CAS RN 1239511-06-8

IUPAC Name: 4-iodo-3,5-bis(trifluoromethyl)-1H-pyrazole
Synonyms: 4-iodo-3,5-bis(trifluoromethyl)-1H-pyrazole
Molecular Weight (g/mol): 329.97
Molecular Formula: C5HF6IN2
InChi Key: OTYORVJCWWESOF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(I)C(=NN1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results