CAS RN 123910-86-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(4-Hydroxy-1-butynyl)benzoic Acid Methyl Ester, TRC
CAS: 123910-86-1 Molecular Formula: C12 H12 O3 Molecular Weight (g/mol): 204.22 Synonym: 4-(4-Hydroxy-1-butyn-1-yl)-benzoic Acid Methyl Ester IUPAC Name: methyl 4-(4-hydroxybut-1-ynyl)benzoate SMILES: COC(=O)c1ccc(cc1)C#CCCO