accessibility menu, dialog, popup

UserName

H3COFOOHF

CAS RN 123843-56-1

IUPAC Name: 4-butoxy-2,6-difluorobenzoic acid
Synonyms: 4-butoxy-2,6-difluorobenzoic acid
Molecular Weight (g/mol): 230.21
Molecular Formula: C11H12F2O3
InChi Key: CSCQGLAJIXARHJ-UHFFFAOYSA-N
SMILES: CCCCOC1=CC(F)=C(C(O)=O)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results