CAS RN 1235034-55-5
CAS RN 1235034-55-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
A-1155463, MedChemExpress
MedChemExpress A-1155463 is a highly potent and selective BCL-XL inhibitor with an EC50 of 70 nM in Molt-4 cell.