CAS RN 1234480-46-6
IUPAC Name:
5-{[2-methoxy-4-(4-methylpiperazin-1-yl)phenyl]amino}-2-methyl-2,4,6,9-tetraazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Synonyms:
5-{[2-methoxy-4-(4-methylpiperazin-1-yl)phenyl]amino}-2-methyl-2,4,6,9-tetraazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Molecular Weight (g/mol):
445.53
Molecular Formula:
C24H27N7O2
InChi Key:
LGLHCXISMKHLIK-UHFFFAOYSA-N
SMILES:
COC1=C(NC2=NC=C3NC(=O)C4=CC=CC=C4N(C)C3=N2)C=CC(=C1)N1CCN(C)CC1
1
–
1
of
1
results
1
XMD8-87, MedChemExpress
MedChemExpress XMD8-87 is a potent TNK2 inhibitor with IC50 values of 38 and 113 nM for the D163E and R806Q mutations, respectively.