accessibility menu, dialog, popup

UserName

CAS RN 1233952-03-8

NFNOO

CAS RN 1233952-03-8

IUPAC Name: 1-(2-fluoro-6-nitrophenyl)pyrrolidine
Synonyms: 1-(2-fluoro-6-nitrophenyl)pyrrolidine
Molecular Weight (g/mol): 210.21
Molecular Formula: C10H11FN2O2
InChi Key: QJQZIIGCNJGTMH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(N2CCCC2)C(F)=CC=C1
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results