accessibility menu, dialog, popup

UserName

CAS RN 1232410-49-9

CH3SOONNONHNH2

CAS RN 1232410-49-9

IUPAC Name: 3-amino-6-(4-methanesulfonylphenyl)-N-phenylpyrazine-2-carboxamide
Synonyms: 3-amino-6-(4-methanesulfonylphenyl)-N-phenylpyrazine-2-carboxamide
Molecular Weight (g/mol): 368.41
Molecular Formula: C18H16N4O3S
InChi Key: DUIHHZKTCSNTGM-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(C=C1)C1=CN=C(N)C(=N1)C(=O)NC1=CC=CC=C1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results