CAS RN 1227923-29-6
CAS RN 1227923-29-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MK-7622, MedChemExpress
MedChemExpress MK-7622 (M1 receptor modulator) is a muscarinic M1 receptor positive allosteric modulator.