CAS RN 122454-29-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-Deethoxymethyl Chlorfenapyr, TRC
CAS: 122454-29-9 Molecular Formula: C12 H5 Br Cl F3 N2 Molecular Weight (g/mol): 349.53 Synonym: 1H-Pyrrole-3-carbonitrile, 4-bromo-2-(4-chlorophenyl)-5-(trifluoromethyl)-,4-Bromo-2-(4-chlorophenyl)-5-(trifluoromethyl)-1H-pyrrole-3-carbonitrile,4-Bromo-2-(4-chlorophenyl)-3-cyano-5-(trifluoromethyl)pyrrole,4-Bromo-2-(p-chlorophenyl)-5-(trifluoromethyl)pyrrole-3-carbonitrile,AC 303268,CL 303268,Econea,Econea 028,Tralopyril IUPAC Name: 4-bromo-2-(4-chlorophenyl)-5-(trifluoromethyl)-1H-pyrrole-3-carbonitrile SMILES: FC(F)(F)c1[nH]c(c2ccc(Cl)cc2)c(C#N)c1Br