CAS RN 1219828-35-9
CAS RN 1219828-35-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
4-Formyl-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]phenyl]benzamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
4-Des[(4-Methyl-1-piperazinyl)methyl]-4-formyl Imatinib, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Des[(4-Methyl-1-piperazinyl)methyl]-4-formyl Imatinib, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
4-Des[(4-Methyl-1-piperazinyl)methyl]-4-formyl Imatinib, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.