CAS RN 1219798-60-3
CAS RN 1219798-60-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(±)-Albuterol-d3 (3-hydroxymethyl-d2; Alpha-d1), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(±)-Albuterol-d3 (3-hydroxymethyl-d2; alpha-d1), CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.