CAS RN 1219795-22-8
CAS RN 1219795-22-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
p-Toluene-d7-sulfonic Acid H2O, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
p-Toluene-d7-sulfonic Acid H2O, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
p-Toluene-d7-sulfonic Acid H2O, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
4
p-Toluene-d7-sulfonic Acid H2O, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.