accessibility menu, dialog, popup

UserName

CH3MgCl

CAS RN 121905-60-0

IUPAC Name: magnesium(2+) 3-methylbenzen-1-ide chloride
Synonyms: magnesium(2+) 3-methylbenzen-1-ide chloride
Molecular Weight (g/mol): 150.89
Molecular Formula: C7H7ClMg
InChi Key: YIDAONIIDXWYSC-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].CC1=C[C-]=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results