CAS RN 1217605-10-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac trans-3'-Hydroxy Cotinine-3-carboxylic Acid, TRC
CAS: 1217605-10-1 Molecular Formula: C11H12N2O4 Molecular Weight (g/mol): 236.22 Synonym: rac trans-3-Hydroxy-1-methyl-2-pyrrolidinone-5-(3-pyridinyl)carboxylic Acid SMILES: Nc1cc(nn1)c2sccc2