CAS RN 121714-22-5
CAS RN 121714-22-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress Fluo-3AM
MedChemExpress Fluo-3AM is a Ca indicator that exhibit increase in fluorescence upon binding Ca2+ used to image spatial dynamics of Ca2+ signaling in FC experiments involving photoactivation of caged chelators, 2nd messengers, and neurotransmitters.