CAS RN 121702-86-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-(-)-Ibuprofen-d3, TRC
CAS: 121702-86-1 Molecular Formula: C13H15D3O2 Molecular Weight (g/mol): 209.3 Synonym: (αR)-α-(Methyl-d3)-4-(2-methylpropyl)benzeneacetic Acid,(-)-Ibuprofen-d3,(R)-2-(4-Isobutylphenyl)propanoic Acid-d3,(R)-Ibuprofen-d3,R-(-)-p-Isobutylhydratropic Acid-d3 SMILES: CC(C)CC1=CC=C([C@@H](C([2H])([2H])[2H])C(O)=O)C=C1