CAS RN 1217-83-0
CAS RN 1217-83-0
IUPAC Name:
2-(4-chlorophenyl)-1H-indole-3-carbaldehyde
Synonyms:
2-(4-chlorophenyl)-1H-indole-3-carbaldehyde
Molecular Weight (g/mol):
255.7
Molecular Formula:
C15H10ClNO
InChi Key:
LISKDSDZIWRRTD-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(C=C1)C1=C(C=O)C2=CC=CC=C2N1