CAS RN 1216840-94-6
CAS RN 1216840-94-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Doxylamine-d5 Succinate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(±)-Doxylamine-d5 Succinate (phenyl-d5), CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
3
Doxylamine-d5 Succinate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.