CAS RN 1216833-74-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Hydroxy Amoxapine-d8, TRC
CAS: 1216833-74-7 Molecular Formula: C17H8D8ClN3O2 Molecular Weight (g/mol): 337.83 Synonym: 2-Chloro-11-(1-piperazinyl-d8)-dibenz[b,f][1,4]oxazepin-7-ol,7-Hydroxyamoxapin-d8 IUPAC Name: 8-chloro-6-(2,2,3,3,5,5,6,6-octadeuteriopiperazin-1-yl)benzo[b][1,4]benzoxazepin-2-ol SMILES: [2H]C1([2H])NC([2H])([2H])C([2H])([2H])N(C2=Nc3ccc(O)cc3Oc4ccc(Cl)cc24)C1([2H])[2H]