CAS RN 1215398-95-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Ethyl-2-phenylmalonamide-d5, TRC
CAS: 1215398-95-0 Molecular Formula: C11 H9 D5 N2 O2 Molecular Weight (g/mol): 211.27 Synonym: PEMA-d5 IUPAC Name: 2-(1,1,2,2,2-pentadeuterioethyl)-2-phenylpropanediamide SMILES: CCC(C1=CC=CC=C1)(C(=O)N)C(=O)N