accessibility menu, dialog, popup

UserName

ReOOOHHH

CAS RN 12079-73-1

IUPAC Name: 1,1,1-tris(hydroxymethyl)-1-rhenapentacyclo[2.2.0.0¹,³.0¹,⁵.0²,⁶]hexan-1-yl
Synonyms: 1,1,1-tris(hydroxymethyl)-1-rhenapentacyclo[2.2.0.0¹,³.0¹,⁵.0²,⁶]hexan-1-yl
Molecular Weight (g/mol): 338.36
Molecular Formula: C8H8O3Re
InChi Key: NEXLWSRPYZNSNH-UHFFFAOYSA-N
SMILES:

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results