CAS RN 120701-87-3
CAS RN 120701-87-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(2S,3S)-2-Amino-3-methyl-3-sulfino-4-(1H-1,2,3-triazol-1-yl)butyric Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.