accessibility menu, dialog, popup

UserName

CAS RN 1206594-24-2

CH3OOHOFFF(E/Z)

CAS RN 1206594-24-2

IUPAC Name: 3-[4-ethoxy-3-(trifluoromethyl)phenyl]prop-2-enoic acid
Synonyms: 3-[4-ethoxy-3-(trifluoromethyl)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 260.21
Molecular Formula: C12H11F3O3
InChi Key: WNOQTODUBHURAA-UHFFFAOYSA-N
SMILES: CCOC1=C(C=C(C=CC(O)=O)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results