CAS RN 1204317-86-1
CAS RN 1204317-86-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Edasalonexent, MedChemExpress
MedChemExpress Edasalonexent (CAT-1004) is an orally bioavailable NF-κB inhibitor.